Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IRF5 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC4508N
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4508N 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4508N Supplier R&D Systems Supplier No. IC4508N
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

IRF5 Polyclonal specifically detects IRF5 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IRF5
Applications Flow Cytometry
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Gene Accession No. NP_002191
Gene Alias IBD14, interferon regulatory factor 5, IRF-5, SLEB10
Gene Symbols IRF5
Host Species Sheep
Immunogen E. coli-derived recombinant human IRF5 Ala214-Thr395 Accession # NP_002191
Purification Method Affinity Purified
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 3663
Test Specificity Detects human IRF5 in Western blots.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.