Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IRF8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5117
This item is not returnable.
View return policy
Description
IRF8 Monoclonal specifically detects IRF8 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
IRF8 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q02556 | |
IRF8 | |
E. coli-derived recombinant human IRF8 Ala255-Val426 Accession # Q02556 | |
100 μg | |
Primary | |
Detects human IRF8 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human IRF4 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
809926 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ICSBP1, ICSBPH-ICSBP, interferon consensus sequence binding protein 1, Interferon consensus sequence-binding protein, interferon regulatory factor 8, IRF-8ICSBP1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3394 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction