Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IRF8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5117SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5117SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5117SP Supplier R&D Systems Supplier No. MAB5117SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IRF8 Monoclonal specifically detects IRF8 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen IRF8
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 809926
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q02556
Gene Alias ICSBP1, ICSBPH-ICSBP, interferon consensus sequence binding protein 1, Interferon consensus sequence-binding protein, interferon regulatory factor 8, IRF-8ICSBP1
Gene Symbols IRF8
Host Species Mouse
Immunogen E. coli-derived recombinant human IRF8 Ala255-Val426 Accession # Q02556
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3394
Test Specificity Detects human IRF8 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human IRF4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.