Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human iRHOM2/RHBDF2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28 - $531.97
Specifications
| Antigen | RHBDF2 |
|---|---|
| Clone | 996308 |
| Dilution | Western Blot 2 μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10048SP
![]() |
R&D Systems
MAB10048SP |
25 μg |
Each for $188.28
|
|
|||||
MAB10048100
![]() |
R&D Systems
MAB10048100 |
100 μg |
Each for $531.97
|
|
|||||
Description
RHBDF2 Monoclonal specifically detects RHBDF2 in Human samples. It is validated for Western Blot.Specifications
| RHBDF2 | |
| Western Blot 2 μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Q6PJF2 | |
| 79651.0 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 996308 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| FLJ22341, iRHOM2, RHBDL5, RHBDL5rhomboid family member 2, RHBDL6, Rhomboid 5 homolog 2, rhomboid 5 homolog 2 (Drosophila), rhomboid veinlet-like 5, Rhomboid veinlet-like protein 5, Rhomboid veinlet-like protein 6, rhomboid, veinlet-like 6, rhomboid, veinlet-like 6 (Drosophila), rhomboid-like protein 6, TOCG | |
| Synthetic peptide containing human iRHOM2/RHBDF2, Accession No. Q6PJF2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects huma iRHOM2/RHBDF2 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title