Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Irisin/FNDC5 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB9420S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9420S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9420S100 Supplier R&D Systems Supplier No. FAB9420S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Irisin/FNDC5 Monoclonal specifically detects Irisin/FNDC5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Irisin/FNDC5
Applications Flow Cytometry
Classification Monoclonal
Clone 936434
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q8NAU1
Gene Alias fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FNDC5, FRCP2, FRCP2Fibronectin type III repeat-containing protein 2
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Irisin/FNDC5, Asp32-Glu143, Accession No. Q8NAU1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 252995.0
Test Specificity Detects human Irisin/FNDC5 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.