Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Irisin/FNDC5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8880100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8880100 100 μg
MAB8880SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8880100 Supplier R&D Systems Supplier No. MAB8880100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Irisin/FNDC5 Monoclonal specifically detects Irisin/FNDC5 in Human samples. It is validated for ELISA.

Specifications

Antigen Irisin/FNDC5
Applications ELISA
Classification Monoclonal
Clone 936410
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q8NAU1
Gene Alias fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FNDC5, FRCP2, FRCP2Fibronectin type III repeat-containing protein 2
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Irisin/FNDC5 Asp32-Glu143 Accession # Q8NAU1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 252995
Test Specificity Detects human Irisin/FNDC5 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.