Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IRS2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6347SP
This item is not returnable.
View return policy
Description
IRS2 Monoclonal specifically detects IRS2 in Human samples. It is validated for Western Blot.Specifications
| IRS2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaN3 with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| insulin receptor substrate 2, IRS-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8660 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 676415 | |
| Western Blot 1 ug/mL | |
| Q9Y4H2 | |
| IRS2 | |
| E. coli-derived recombinant human IRS2 Pro1035-Ile1136 Accession # Q9Y4H2 | |
| 25 μg | |
| Primary | |
| Detects human IRS2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human IRS1 or recombinant mouse IRS3 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction