Learn More
Human ISLR-2 Antibody, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody
Supplier:  R&D Systems AF4526
 View more versions of this product
Catalog No. AF4526
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
ISLR-2 Polyclonal specifically detects ISLR-2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
ISLR-2
Polyclonal
Unconjugated
Q6UXK2
ISLR2
Mouse myeloma cell line NS0-derived recombinant human ISLR-2 Cys20-Leu590 Accession # Q6UXK2
100 μg
Primary
Detects human ISLR-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human ISLR-2 is observed.
Human
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
immunoglobulin superfamily containing leucine-rich repeat 2, ISLR2, KIAA1465, leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein, LINX, UNQ1885/PRO4329
Sheep
Affinity Purified
RUO
57611
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only