Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ITIH4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8157
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8157 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8157 Supplier R&D Systems Supplier No. AF8157
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ITIH4 Polyclonal specifically detects ITIH4 in Human samples. It is validated for Western Blot.

Specifications

Antigen ITIH4
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q14624
Gene Alias DKFZp686G21125, gp120, H4P, heavy chain-like, 1, IHRP, inter-alpha (globulin) inhibitor H4 (plasma Kallikrein-sensitive glycoprotein), inter-alpha-trypsin inhibitor family heavy chain-related protein, inter-alpha-trypsin inhibitor heavy chain H4, ITI-HC4, ITIHL1, PK120, PK-120, plasma kallikrein-sensitive glycoprotein 120, PRO1851
Gene Symbols ITIH4
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ITIH4 Glu29-Leu930 (Ile85Asn, Pro698Thr) Accession # Q14624
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3700
Test Specificity Detects human ITIH4 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human ITIH1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.