Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ITM2A Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4876
This item is not returnable.
View return policy
Description
ITM2A Polyclonal specifically detects ITM2A in Human samples. It is validated for Western Blot.Specifications
ITM2A | |
Polyclonal | |
Unconjugated | |
O43736 | |
ITM2A | |
E. coli-derived recombinant human ITM2A Pro80-Glu263 Accession # O43736 | |
100 μg | |
Primary | |
Detects human ITM2A in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) ITM2B is observed, and less than 1% cross-reactivity with rhITM2C is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
BRICD2A, BRICHOS domain containing 2A, E25A, E25ABRICD2A, integral membrane protein 2A, Protein E25 | |
Sheep | |
Affinity Purified | |
RUO | |
9452 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction