Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ITM2B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7015
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7015 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7015 Supplier R&D Systems Supplier No. AF7015
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ITM2B Polyclonal specifically detects ITM2B in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen ITM2B
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q9Y287
Gene Alias ABRI, ABri/ADan amyloid peptide, BRI, BRI2, BRICD2B, BRICHOS domain containing 2B, BRIFBD, E25B, E3-16, FBD, integral membrane protein 2B, Protein E25B, Transmembrane protein BRI
Gene Symbols ITM2B
Host Species Sheep
Immunogen E. coli-derived recombinant human ITM2B Ala60-Ser266 Accession # Q9Y287
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9445
Test Specificity Detects human ITM2B in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ITM2A and rhITM2C is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.