Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ITM2C Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4904
This item is not returnable.
View return policy
Description
ITM2C Polyclonal specifically detects ITM2C in Human samples. It is validated for Western Blot.Specifications
ITM2C | |
Polyclonal | |
Unconjugated | |
Q9NQX7 | |
ITM2C | |
E. coli-derived recombinant human ITM2C Ala86-Val267 Accession # Q9NQX7 | |
100 μg | |
Primary | |
Detects human ITM2C in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) ITM2A and rhITM2B is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
BRI3, BRI3E25C, BRICD2C, BRICHOS domain containing 2C, Cerebral protein 14, E25, hRPC.1050_D_4, integral membrane protein 2C, integral membrane protein 3, ITM3, Transmembrane protein BRI3 | |
Sheep | |
Affinity Purified | |
RUO | |
81618 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction