Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IVL (P07476, 420 a.a. - 578 a.a.) Partial Recombinant Protein

Catalog No. 89949381
Click to view available options
:
10μg

Used for SDS-PAGE

Involucrin, a component of the keratinocyte crosslinked envelope, is found in the cytoplasm and crosslinked to membrane proteins by transglutaminase. This gene is mapped to 1q21, among calpactin I light chain, trichohyalin, profillaggrin, loricrin, and calcyclin. [provided by RefSeq]

Specifications

Accession Number P07476
Concentration 0.1mg/mL
For Use With (Application) SDS-PAGE
Formulation In 50mM Tris-HCl, 10mM GSH, pH 7.5
Gene ID (Entrez) 3713
Molecular Weight (g/mol) 41kDa
Name Human IVL (P07476, 420 a.a. - 578 a.a.) Partial Recombinant Protein
Quantity 10μg
Storage Requirements Store at −80°C.Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias NY-ESO-1
Common Name IVL
Gene Symbol IVL
Species E. coli
Recombinant Recombinant
Protein Tag Untagged
Expression System Escherichia coli expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.