Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Jade-1/PHF17 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6275
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6275 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6275 Supplier R&D Systems Supplier No. MAB6275
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Jade-1/PHF17 Monoclonal specifically detects Jade-1/PHF17 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Jade-1/PHF17
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 658005
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6IE81
Gene Alias FLJ22479, Jade1, KIAA1807, PHD finger protein 17JADE1gene for apoptosis and differentiation in epithelia, PHD protein Jade-1, PHF17, protein Jade-1
Gene Symbols PHF17
Host Species Mouse
Immunogen E. coli-derived recombinant human JADE-1/PHF-17 Met1-Ser137 Accession # Q6IE81
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79960
Test Specificity Detects human JADE-1/PHF-17 in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human Jade-2/PHF15 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.