Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Jagged 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $1973.00
Specifications
Antigen | Jagged 1 |
---|---|
Clone | 188323 |
Concentration | LYOPH |
Dilution | Neutralization 0.5-3 ug/mL |
Applications | Neutralization |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB12771SP
![]() |
R&D Systems
MAB12771SP |
25 μg |
Each for $200.50
|
|
|||||
MAB12771100
![]() |
R&D Systems
MAB12771100 |
100 μg |
Each for $561.00
|
|
|||||
MAB12771500
![]() |
R&D Systems
MAB12771500 |
500 μg |
Each for $1,973.00
|
|
|||||
Description
Jagged 1 Monoclonal specifically detects Jagged 1 in Human samples. It is validated for Neutralization.Specifications
Jagged 1 | |
LYOPH | |
Neutralization | |
Unconjugated | |
Mouse | |
Human | |
P78504 | |
182 | |
NS0-derived recombinant human Jagged 1 Ser32-Asp296 Accession # P78504 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Jagged 1 in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant human Jagged 2 and recombinant rat Jagged 1 is observed. |
188323 | |
Neutralization 0.5-3 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, JAG1, jagged 1, Jagged1, JAGL1, protein jagged-1 | |
JAG1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title