Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Jagged 2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1726
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1726 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1726 Supplier R&D Systems Supplier No. BAF1726
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Jagged 2 Polyclonal specifically detects Jagged 2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Jagged 2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9Y219
Gene Alias HJ2, JAG2, jagged 2, Jagged2, protein jagged-2, SER2
Gene Symbols JAG2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Jagged 2 Met27-Asp307 Accession # Q9Y219
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3714
Test Specificity Detects human Jagged 2 in Western blots. In Western blots, approximately 35% cross-reactivity with recombinant human Jagged 1 is observed and less than 1% cross-reactivity with recombinant rat Jagged 1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.