Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Jak3 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC46991G100UG
This item is not returnable.
View return policy
Description
Jak3 Monoclonal specifically detects Jak3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Jak3 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
EC 2.7.10, JAK3_HUMAN, JAK-3EC 2.7.10.2, JAKL, JAKLtyrosine-protein kinase JAK3, Janus kinase 3Leukocyte janus kinase, LJAK, L-JAKJanus kinase 3 (a protein tyrosine kinase, leukocyte) | |
E. coli-derived recombinant human Jak3 Gly46-Thr209 Accession # P52333 | |
RUO | |
3718 | |
Human | |
IgG2b |
Flow Cytometry | |
452524 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P52333 | |
Mouse | |
100 μg | |
Primary | |
Detects human Jak3 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction