Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Jak3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4699SP
This item is not returnable.
View return policy
Description
Jak3 Monoclonal specifically detects Jak3 in Human samples. It is validated for Western Blot.Specifications
Jak3 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
P52333 | |
JAK3 | |
E. coli-derived recombinant human Jak3 Gly46-Thr209 Accession # P52333 | |
25 μg | |
Primary | |
Detects human Jak3 in Western blots. | |
Human | |
Purified |
Western Blot | |
452506 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, JAK3_HUMAN, JAK-3EC 2.7.10.2, JAKL, JAKLtyrosine-protein kinase JAK3, Janus kinase 3Leukocyte janus kinase, LJAK, L-JAKJanus kinase 3 (a protein tyrosine kinase, leukocyte) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3718 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction