Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Jak3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB46991
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB46991 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB46991 Supplier R&D Systems Supplier No. MAB46991
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Jak3 Monoclonal specifically detects Jak3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen Jak3
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 452524
Concentration LYOPH
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P52333
Gene Alias EC 2.7.10, JAK3_HUMAN, JAK-3EC 2.7.10.2, JAKL, JAKLtyrosine-protein kinase JAK3, Janus kinase 3Leukocyte janus kinase, LJAK, L-JAKJanus kinase 3 (a protein tyrosine kinase, leukocyte)
Gene Symbols JAK3
Host Species Mouse
Immunogen E. coli-derived recombinant human Jak3 Gly46-Thr209 Accession # P52333
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3718
Test Specificity Detects human Jak3 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.