Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human JAM-A Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1103G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1103G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1103G100 Supplier R&D Systems Supplier No. FAB1103G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

JAM-A Monoclonal specifically detects JAM-A in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen JAM-A
Applications Flow Cytometry
Classification Monoclonal
Clone 654806
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9Y624
Gene Alias CD321, CD321 antigen, F11 receptor, F11R, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1, PAM-1KAT, Platelet adhesion molecule 1
Gene Symbols F11R
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human JAM-A, Ser28-Ala242 (predicted), Accession # Q9Y624
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 50848
Test Specificity Detects human JAM-A in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) JAM-B, recombinant mouse (rm) JAM-A, or rmJAM-4 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.