Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human JAM-A/CD321/F11R ELISA Kit

Catalog No. p-7219477
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests

ELISA

Human JAM-A/CD321/F11R quantitates human JAM-A/CD321/F11R in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human JAM-A/CD321/F11R.

CD321 (F11R, JAM-1, JAM-A) is a cell-surface adhesion molecule belonging to the Ig superfamily of receptors. CD321 is a 32-41 kD glycoprotein, possessing two extracellular Ig-like domains, a transmembrane region, and a short cytosolic tail. The protein assembles at the membrane as a homodimer, and can associate laterally with other membrane proteins including integrins. CD321 is expressed on a variety of cell types, including leukocytes, platelets, erythrocytes, stem cells, epithelial and endothelial cells, and plays a role in mediating cell-cell contact. CD321 can be found in close proximity to several components of epithelial tight junctions, including ZO-1 and occludin. CD321 is involved in the maintenance of epithelial and endothelial barriers, hematopoiesis, angiogenesis, and tumor proliferation, invasion, and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 9130004G24; AA638916; BV11 antigen; CD321; ESTM33; F11 R; F11 receptor; F11 receptor/F11R; F11r; JAM; Jam1; JAM-1; JAMA; JAM-A; Jcam; Jcam1; junction cell adhesion molecule A; junction cell adhesion molecule1; junctional adhesion molecule 1; Junctional adhesion molecule A; KAT; Ly106; PAM-1; Platelet adhesion molecule 1; platelet F11 receptor; UNQ264/PRO301
Assay Range 13 to 10,000 pg/mL
Assay Sensitivity 13 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 50848
Gene Symbol F11r
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target JAM-A (CD321)
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.