Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human JAM-B/VE-JAM Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA10742V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA10742V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA10742V100 Supplier R&D Systems Supplier No. FAB10742V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

JAM-B/VE-JAM Monoclonal specifically detects JAM-B/VE-JAM in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen JAM-B/VE-JAM
Applications Flow Cytometry
Classification Monoclonal
Clone 988901
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 μg/10^6 cells
Formulation Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P57087
Gene Alias C21orf43, CD322, CD322 antigen, JAM2, JAMB, JAM-BJAM-IT/VE-JAM, junctional adhesion molecule 2JAM-2, junctional adhesion molecule B, PRO245, Vascular endothelial junction-associated molecule, VE-JAM, VE-JAMVEJAMCD322
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human JAM-B/VE-JAM Phe29-Asn236 Accession # P57087
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 58494
Test Specificity Detects human JAM-B/VE-JAM in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.