Learn More
Human JAM-B/VE-JAM Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB10743R100UG
Catalog No. FB10743R100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
JAM-B/VE-JAM Monoclonal specifically detects JAM-B/VE-JAM in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
JAM-B/VE-JAM
Monoclonal
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
C21orf43, CD322, CD322 antigen, JAM2, JAMB, JAM-BJAM-IT/VE-JAM, junctional adhesion molecule 2JAM-2, junctional adhesion molecule B, PRO245, Vascular endothelial junction-associated molecule, VE-JAM, VE-JAMVEJAMCD322
Mouse myeloma cell line NS0-derived recombinant human JAM-B/VE-JAM, Phe29-Asn236, Accession No. P57087
100 μg
58494.0
Human
Supernatant
Flow Cytometry
988934
Flow Cytometry 0.25-1 ug/10^6 cells
P57087
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human JAM-B/VE-JAM in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.