Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human JAM-C Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF1189
This item is not returnable.
View return policy
Description
JAM-C Polyclonal specifically detects JAM-C in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Neutralization, CyTOF-ready.Specifications
JAM-C | |
Polyclonal | |
Unconjugated | |
Q9BX67 | |
JAM3 | |
Mouse myeloma cell line NS0-derived recombinant human JAM-C Val32-Asn241 (Ala149Pro) Accession # Q9BX67 | |
100 μg | |
Primary | |
Detects human JAM-C in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 75% cross-reactivity with recombinant mouse (rm) JAM-C is observed and less than 1% cross-reactivity with recombinant human JAM-1 and rmJAM-1 is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, Neutralization, CyTOF | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Neutralization 2-8 ug/mL, CyTOF-ready | |
CD323, JAM-2, JAM3, JAMC, JAM-CFLJ14529, junctional adhesion molecule 3JAM-3, junctional adhesion molecule C | |
Goat | |
Affinity Purified | |
RUO | |
83700 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction