Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human JARID2 Partial ORF (NP_004964, 1130 a.a. - 1229 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89949416
Click to view available options
Quantity:
10 ug
25 ug

Used for AP, Array, ELISA, WB-Re

This gene is an ortholog of the mouse jumonji gene, which encodes a nuclear protein essential for mouse embryogenesis, including neural tube formation. Overexpression of mouse jumonji negatively regulates cell proliferation. The jumonji proteins contain a DNA-binding domain, called an AT-rich interaction domain (ARID), and share regions of similarity with human retinoblastoma-binding protein-2 and the human SMCX protein. [provided by RefSeq]

Sequence: LQLETSERRCQICQHLCYLSMVVQENENVVFCLECALRHVEKQKSCRGLKLMYRYDEEQIISLVNQICGKVSGKNGSIENCLSKPTPKRGPRKRATVDVP

Specifications

Accession Number NP_004964
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 3720
Molecular Weight (g/mol) 36.74kDa
Name JARID2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen LQLETSERRCQICQHLCYLSMVVQENENVVFCLECALRHVEKQKSCRGLKLMYRYDEEQIISLVNQICGKVSGKNGSIENCLSKPTPKRGPRKRATVDVP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias JMJ
Common Name JARID2
Gene Symbol JARID2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.