Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human JunB Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4456
This item is not returnable.
View return policy
Description
JunB/AP-1 Monoclonal specifically detects JunB/AP-1 in Human samples. It is validated for Western Blot.Specifications
| JunB/AP-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P17275 | |
| JUNB | |
| E. coli-drived recombinant human JunB Asp145-Val246 (predicted) Accession # P17275 | |
| 100 μg | |
| Primary | |
| Detects human JunB in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) JunD or rhc-Jun is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 512313 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| activator protein 1, AP-1, jun B proto-oncogene, transcription factor jun-B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3726 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction