Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KA1/GRIK4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB11326100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11326100 100 μg
MAB11326SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB11326100 Supplier R&D Systems Supplier No. MAB11326100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KA1/GRIK4/Glutamate Receptor KA1 Monoclonal antibody specifically detects KA1/GRIK4/Glutamate Receptor KA1 in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen KA1/GRIK4/Glutamate Receptor KA1
Applications Immunohistochemistry
Classification Monoclonal
Clone 1060003
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. P42263
Gene Alias EAA1, Excitatory amino acid receptor 1, GluK4, glutamate receptor KA1, Glutamate receptor KA-1, glutamate receptor, ionotropic kainate 4, glutamate receptor, ionotropic, kainate 4, GRIK4, KA1GRIK
Host Species Mouse
Immunogen Human KA1/GRIK containing peptide, Accession # P42263
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2900
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.