Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KA2/GRIK5 Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB11334SP
This item is not returnable.
View return policy
Description
KA2/GRIK5/Glutamate Receptor KA2 Monoclonal antibody specifically detects KA2/GRIK5/Glutamate Receptor KA2 in Human samples. It is validated for ImmunohistochemistrySpecifications
KA2/GRIK5/Glutamate Receptor KA2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
EAA2, Excitatory amino acid receptor 2, GluK5, glutamate receptor KA2, Glutamate receptor KA-2, glutamate receptor, ionotropic kainate 5, glutamate receptor, ionotropic, kainate 5, GRIK2, GRIK5, KA2GRIK2 | |
Synthetic peptide sequence, Accession # Q16478 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2b |
Immunohistochemistry | |
1060109 | |
Immunohistochemistry 5-25 ug/mL | |
Q16478 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2901 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction