Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KA2/GRIK5 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$172.81 - $459.11
Specifications
| Antigen | KA2/GRIK5/Glutamate Receptor KA2 |
|---|---|
| Clone | 2916E |
| Dilution | Immunohistochemistry 5-15 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB113341SP
![]() |
R&D Systems
MAB113341SP |
25 μg |
Each for $172.81
|
|
|||||
RD10000183
![]() |
R&D Systems
MAB113341100 |
100 μg |
Each for $459.11
|
|
|||||
Description
KA2/GRIK5/Glutamate Receptor KA2 Monoclonal antibody specifically detects KA2/GRIK5/Glutamate Receptor KA2 in Human samples. It is validated for ImmunohistochemistrySpecifications
| KA2/GRIK5/Glutamate Receptor KA2 | |
| Immunohistochemistry 5-15 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| EAA2, Excitatory amino acid receptor 2, GluK5, glutamate receptor KA2, Glutamate receptor KA-2, glutamate receptor, ionotropic kainate 5, glutamate receptor, ionotropic, kainate 5, GRIK2, GRIK5, KA2GRIK2 | |
| Synthetic peptide sequence, Accession # Q16478 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2916E | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q16478 | |
| 2901 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title