Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Kallikrein 11 His-tag Recombinant Protein

Catalog No. PIRP88087
Encompass
Click to view available options
Quantity:
50 μg

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (250 μL) be added to the vial to prepare a stock solution of 0.2 mg/mL. Concentration is measured by UV.

Hippostasin, also known as KLK11, is a kallikrein-like serine protease, which has two alternatively spliced isoforms, brain-type and prostate-type. Hippostasin plays a role in the prostate, including reproductive and/or tumorigenic functions. Elevated serum levels of KLK11 were found in 70% of women with ovarian cancer and in 60% of men with prostate cancer. Analysis of the KLK11 biomarker in serum may aid in the diagnosis and monitoring of ovarian and prostatic carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UBX7
For Use With (Application) Functional Assay
Format Lyophilized
Formulation PBS with 0.01% mannitol, 0.01% Tween 80, 5-8% trehalose and no preservative; pH 7.4
Gene ID (Entrez) 11012
Molecular Weight (g/mol) 40 kDa
Name Human Kallikrein 11 His-tag
Purification Method SDS-PAGE
Quantity 50 μg
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias 2310015I08Rik; Hippostasin; hK11; kallikrein 11; kallikrein related peptidase 11; kallikrein related-peptidase 11; Kallikrein11; kallikrein-11; Kallikrein-11 inactive chain 1; Kallikrein-11 inactive chain 2; kallikrein-related peptidase 11; KLK11; protease, serine, 20; Prss20; Serine protease 20; TLSP; Trypsin-like protease; UNQ649/PRO1279
Common Name Kallikrein 11
Gene Symbol Klk11
Biological Activity Measured by its ability to cleave a colorimetric peptide substrate D-Val-Leu-Lys-ThioBenzyl ester (VLK-SBzl), in the presence of 5,5'Dithio-bis (2-nitrobenzoic acid) (DTNB) (Edwards, K.M. et al.,1999, J. Biol. Chem. 274: 30468). The specific activity is >200 pmoles/min/ μg. (Activation description: The proenzyme needs to be activated by Thermolysin for an activated form)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence Human KLK11 isoform 1, amino acids Met1-Asn250 (Accession # NP_006844.1) with a C-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Measured by its ability to cleave a colorimetric peptide substrate D-Val-Leu-Lys-ThioBenzyl ester (VLK-SBzl), in the presence of 5,5'Dithio-bis (2-nitrobenzoic acid) (DTNB) (Edwards, K.M. et al.,1999, J. Biol. Chem. 274: 30468). The specific activity is >200 pmoles/min/ μg. (Activation description: The proenzyme needs to be activated by Thermolysin for an activated form)
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.