Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Kallikrein 13 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2625SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2625SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2625SP Supplier R&D Systems Supplier No. AF2625SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Kallikrein 13 Polyclonal specifically detects Kallikrein 13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Neutralization.

Specifications

Antigen Kallikrein 13
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 8.6 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UKR3
Gene Alias DKFZp586J1923, EC 3.4.21, EC 3.4.21.-, kallikrein 13, Kallikrein-like protein 4, kallikrein-related peptidase 13, KLK13, KLK-L4kallikrein-13, KLKL4kallikrein-like gene 4
Gene Symbols KLK13
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kallikrein 13 Val19-Ile262 Accession # Q9UKR3
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26085
Test Specificity Detects human Kallikrein 13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) Kallikrein 8 and less than 5% cross-reactivity with rhKallikrein 11, rhKallikrein 14, and rhKallikrein 15 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.