Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kallikrein 13 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | Kallikrein 13 |
---|---|
Clone | 341419 |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 2.4 ug/mL |
Applications | Western Blot, Immunoprecipitation, Neutralization |
Classification | Monoclonal |
Description
Kallikrein 13 Monoclonal specifically detects Kallikrein 13 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
Kallikrein 13 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 2.4 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp586J1923, EC 3.4.21, EC 3.4.21.-, kallikrein 13, Kallikrein-like protein 4, kallikrein-related peptidase 13, KLK13, KLK-L4kallikrein-13, KLKL4kallikrein-like gene 4 | |
KLK13 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
341419 | |
Western Blot, Immunoprecipitation, Neutralization | |
Unconjugated | |
Mouse | |
Human | |
Q9UKR3 | |
26085 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 13 Val19-Ile262 Accession # Q9UKR3 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Kallikrein 13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Kallikrein 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 14, 15, rhKallikrein B1, rhFactorVII, rhHGFA, rhUPA, rhThrombin, rhFactor X, or rhFactor XI is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title