Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kallikrein 3/PSA Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
| Antigen | Kallikrein3/PSA |
|---|---|
| Clone | 989729 |
| Dilution | Western Blot 2μg/mL, Immunohistochemistry 5-25μg/mL, Immunocytochemistry 5-25μg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB13443-SP
![]() |
R&D Systems
MAB13443SP |
25 μg |
Each for $200.50
|
|
|||||
MAB13443-100
![]() |
R&D Systems
MAB13443100 |
100 μg |
Each for $502.50
|
|
|||||
Description
Kallikrein 3/PSA Monoclonal specifically detects Kallikrein 3/PSA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| Kallikrein3/PSA | |
| Western Blot 2μg/mL, Immunohistochemistry 5-25μg/mL, Immunocytochemistry 5-25μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P07288 | |
| 354 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 989729 | |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, KLK3, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 3/PSA Pro19-Pro261 Accession # P07288 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human Kallikrein 3/PSA in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title