Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kallikrein 3/PSA Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF1344
This item is not returnable.
View return policy
Description
Kallikrein 3/PSA Polyclonal specifically detects Kallikrein 3/PSA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.Specifications
Kallikrein 3/PSA | |
Polyclonal | |
Unconjugated | |
P07288 | |
KLK3 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 3 Ala18-Pro261 Accession # P07288 | |
100 μg | |
Primary | |
Detects human KLK3 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) KLK2 is observed and less than 5% cross-reactivity with rhKLK1, rhKLK4, rhKLK5, rhKLK7, rhKLK8, rhKLK9, rhKLK10, rhKLK11, rhKLK12, rhKLK13, and rhKLK15 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
LYOPH | |
Western Blot 0.5-1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, KLK3, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin | |
Goat | |
Affinity Purified | |
RUO | |
354 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction