Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kallikrein 4/Prostase/EMSP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB35661
This item is not returnable.
View return policy
Description
Kallikrein 4/Prostase/EMSP1 Monoclonal specifically detects Kallikrein 4/Prostase/EMSP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Neutralization.Specifications
Kallikrein 4/Prostase/EMSP1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AI2A1, ARM1, EC 3.4.21, EC 3.4.21.-, EMSP, EMSP1, EMSP1MGC116827, Enamel matrix serine proteinase 1, kallikrein 4 (prostase, enamel matrix, prostate), kallikrein-4, Kallikrein-like protein 1, kallikrein-related peptidase 4, KLK4, KLK-L1MGC116828, Prostase, PRSS17enamel matrix serine protease 1, PSTSandrogen-regulated message 1, Serine protease 17 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9622 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunoprecipitation, Neutralization | |
325712 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 2.9 ug/mL | |
NP_004908 | |
KLK4 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 4/Prostase/EMSP1 Ser27-Ser254 Accession # NP_004908 | |
500 μg | |
Primary | |
Detects human Kallikrein 4/Prostase/EMSP1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Kallikrein 1, 3, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, 15, rhKallikrein B1, rhFactor VII, rhFactor X, rhFactor XI, rhHGFA, rhThrombin, and rhUPA-1 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction