Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kallikrein 5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1108
This item is not returnable.
View return policy
Description
Kallikrein 5 Polyclonal specifically detects Kallikrein 5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
Kallikrein 5 | |
Polyclonal | |
Unconjugated | |
Q9Y337 | |
KLK5 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 5 Ile67-Ser293 Accession # Q9Y337 | |
100 μg | |
Primary | |
Detects human Kallikrein 5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) KLK8 and rhKLK13 is observed and less than 1% cross-reactivity with rhKLK1, -3, -4, -7, -9, -10, -11, and -12 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
EC 3.4.21, EC 3.4.21.4, kallikrein 5, Kallikrein-like protein 2, kallikrein-related peptidase 5, KLK5, KLKL2, KLK-L2, KLK-L2EC 3.4.21.-, SCTE, SCTEkallikrein-5, Stratum corneum tryptic enzyme | |
Goat | |
Affinity Purified | |
RUO | |
25818 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction