Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kallikrein 5 Propeptide Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $503.00
Specifications
Antigen | Kallikrein 5 |
---|---|
Clone | 325302 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11082SP
![]() |
R&D Systems
MAB11082SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11082
![]() |
R&D Systems
MAB11082 |
100 μg |
Each for $503.00
|
|
|||||
Description
Kallikrein 5 Monoclonal specifically detects Kallikrein 5 in Human samples. It is validated for Western Blot.Specifications
Kallikrein 5 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
NP_036559 | |
25818 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 5 Val23-Arg66 Accession # NP_036559 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Kallikrein 5 Propeptide in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Kallikrein 5 (mature), rhKallikrein 1, -3, -4, -6, -7, -8, -9, -10, -11, -12, -13, -14, -15, rhKallikrein B1, rhHGFA, rhFactor VII, rhFactor X, rhFactor XI, rhThrombin, or rhuPA-1 is observed. |
325302 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.21, EC 3.4.21.4, kallikrein 5, Kallikrein-like protein 2, kallikrein-related peptidase 5, KLK5, KLKL2, KLK-L2, KLK-L2EC 3.4.21.-, SCTE, SCTEkallikrein-5, Stratum corneum tryptic enzyme | |
KLK5 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title