Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kallikrein 7 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$200.50 - $667.00
Specifications
Antigen | Kallikrein 7 |
---|---|
Clone | 333931 |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 5.5 ug/mL |
Applications | Western Blot, Immunoprecipitation, Neutralization |
Classification | Monoclonal |
Description
Kallikrein 7 Monoclonal specifically detects Kallikrein 7 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
Kallikrein 7 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 5.5 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.21, EC 3.4.21.117, hK7, hSCCE, kallikrein 7 (chymotryptic, stratum corneum), kallikrein-related peptidase 7, KLK7, protease, serine, 6, PRSS6, SCCEkallikrein-7, Serine protease 6, signal protein, Stratum corneum chymotryptic enzyme | |
KLK7 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
333931 | |
Western Blot, Immunoprecipitation, Neutralization | |
Unconjugated | |
Mouse | |
Human | |
P49862 | |
5650 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 7 Glu23-His252 Accession # P49862 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Kallikrein 7 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Kallikrein 1, 3, 4, 5, 6, 8, 9, 10, 11, 12, 13, 14, or 15 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title