Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kallikrein 8/Neuropsin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $549.50
Specifications
Antigen | Kallikrein 8/Neuropsin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Neutralization 8 ug/mL |
Applications | Western Blot, Neutralization |
Classification | Polyclonal |
Description
Kallikrein 8/Neuropsin Polyclonal specifically detects Kallikrein 8/Neuropsin in Human samples. It is validated for Western Blot, Neutralization.Specifications
Kallikrein 8/Neuropsin | |
Western Blot 0.1 ug/mL, Neutralization 8 ug/mL | |
Polyclonal | |
Goat | |
Human | |
EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19, PRSS19kallikrein-8, Serine protease 19, Serine protease TADG-14, TADG14neuropsin, Tumor-associated differentially expressed gene 14 protein | |
KLK8 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
O60259 | |
11202 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 8 Gln29-Gly260 Accession # O60259 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Kallikrein 8/Neuropsin in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title