Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kallikrein 8/Neuropsin Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | Kallikrein 8/Neuropsin |
---|---|
Clone | 369910 |
Concentration | LYOPH |
Dilution | Immunoprecipitation 25 ug/mL, Neutralization 0.7 ug/mL |
Applications | Immunoprecipitation, Neutralization |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB17191SP
![]() |
R&D Systems
MAB17191SP |
25 μg |
Each for $200.50
|
|
|||||
MAB17191
![]() |
R&D Systems
MAB17191 |
100 μg |
Each for $468.00
|
|
|||||
Description
Kallikrein 8/Neuropsin Monoclonal specifically detects Kallikrein 8/Neuropsin in Human samples. It is validated for Immunoprecipitation, Neutralization.Specifications
Kallikrein 8/Neuropsin | |
LYOPH | |
Immunoprecipitation, Neutralization | |
Unconjugated | |
Rat | |
Human | |
O60259 | |
11202 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 8/Neuropsin Gln29-Gly260 (predicted) Accession # O60259 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Kallikrein 8/Neuropsin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) UPA-1, rhKallikrein 1, 2, 3, 4, 5, 7, 9, 10, 11, 12, 13, 14, 15, rhKallikrein B1, rhHGFA, rhFactor VII, rhFactor X, rhFactor XI, rhProc, or rhThrombin is observed. |
369910 | |
Immunoprecipitation 25 ug/mL, Neutralization 0.7 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19, PRSS19kallikrein-8, Serine protease 19, Serine protease TADG-14, TADG14neuropsin, Tumor-associated differentially expressed gene 14 protein | |
KLK8 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title