Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kallikrein 8/Neuropsin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1719
This item is not returnable.
View return policy
Description
Kallikrein 8/Neuropsin Monoclonal specifically detects Kallikrein 8/Neuropsin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
Kallikrein 8/Neuropsin | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19, PRSS19kallikrein-8, Serine protease 19, Serine protease TADG-14, TADG14neuropsin, Tumor-associated differentially expressed gene 14 protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
11202 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
189004 | |
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 1-25 ug/mL | |
O60259 | |
KLK8 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 8/Neuropsin Gln29-Gly260 (predicted) Accession # O60259 | |
500 μg | |
Primary | |
Detects human Kallikrein 8/Neuropsin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Kallikrein 3, 5, 11, or rhHGFA is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction