Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kallikrein 9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
| Antigen | Kallikrein 9 |
|---|---|
| Clone | 366721 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
Description
Kallikrein 9 Monoclonal specifically detects Kallikrein 9 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Kallikrein 9 | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UKQ9 | |
| 284366 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 9 Asp20-Asn250 Accession # Q9UKQ9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Kallikrein 9 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Kallikrein 1, 3, 4, 5, 6, 7, 8, 10, 11, 12, 13, 14, 15, rhKLKB1, rhHGFA, rhFactor VII, rhFactor X, rhFactor XI, or rhThrombin is observed. |
| 366721 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, kallikrein 9, Kallikrein-like protein 3, kallikrein-related peptidase 9, KLK9, KLKL3, KLK-L3, KLK-L3kallikrein-9, MGC50513 | |
| KLK9 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title