Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Katanin p60 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7100SP
This item is not returnable.
View return policy
Description
Katanin p60 Monoclonal specifically detects Katanin p60 in Human samples. It is validated for Immunohistochemistry.Specifications
| Katanin p60 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| O75449 | |
| KATNA1 | |
| E. coli-derived recombinant human Katanin p60 Asp308-Tyr378 Accession # O75449 | |
| 25 μg | |
| Primary | |
| Detects human Katanin p60 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Katanin p60 subunit A-like 2 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 717705 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.6.4.3, katanin p60 (ATPase containing) subunit A 1, katanin p60 (ATPase-containing) subunit A 1, katanin p60 ATPase-containing subunit A1, Katanin p60 subunit A1, KATNA1, p60 katanin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11104 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction