Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KCC2/SLC12A5 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8369G100UG
This item is not returnable.
View return policy
Description
KCC2/SLC12A5 Monoclonal specifically detects KCC2/SLC12A5 in Human samples. It is validated for Flow Cytometry.Specifications
KCC2/SLC12A5 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9H2X9 | |
SLC12A5 | |
NS0 mouse myeloma cell line transfected with human SLC12A5 Accession # Q9H2X9 | |
RUO | |
57468 | |
Human | |
IgG1 |
Flow Cytometry | |
905705 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Electroneutral potassium-chloride cotransporter 2, hKCC2, KCC2K-Cl cotransporter 2, KIAA1176erythroid K-Cl cotransporter 2, Neuronal K-Cl cotransporter, SLC12A5, solute carrier family 12 (potassium/chloride transporter), member 5, solute carrier family 12 member 5 | |
Mouse | |
100 μg | |
Primary | |
Detects human SLC12A5 in ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction