Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KCNAB3 Full-length ORF (NP_004723.2, 1 a.a. - 404 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89949506
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. This member is one of the beta subunits, which are auxiliary proteins associating with functional Kv-alpha subunits. This member and the KCNA5 gene product assemble into a heteromultimeric A-type channel that inactivates completely and is significantly faster than other A-type Kv channels. [provided by RefSeq]

Sequence: MQVSIACTEQNLRSRSSEDRLCGPRPGPGGGNGGPAGGGHGNPPGGGGSGPKARAALVPRPPAPAGALRESTGRGTGMKYRNLGKSGLRVSCLGLGTWVTFGSQISDETAEDVLTVAYEHGVNLFDTAEVYAAGKAERTLGNILKSKGWRRSSYVITTKIFWGGQAETERGLSRKHIIEGLRGSLERLQLGYVDIVFANRSDPNCPMEEIVRAMTYVINQGLALYWGTSRWGAAEIMEAYSMARQFNLIPPVCEQAEHHLFQREKVEMQLPELYHKIGVGSVTWYPLACGLITSKYDGRVPDTCRASIKGYQWLKDKVQSEDGKKQQAKVMDLLPVAHQLGCTVAQLAIAWCLRSEGVSSVLLGVSSAEQLIEHLGALQVLSQLTPQTVMEIDGLLGNKPHSKK

Specifications

Accession Number NP_004723.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9196
Molecular Weight (g/mol) 70.1kDa
Name KCNAB3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 25 μg
Immunogen MQVSIACTEQNLRSRSSEDRLCGPRPGPGGGNGGPAGGGHGNPPGGGGSGPKARAALVPRPPAPAGALRESTGRGTGMKYRNLGKSGLRVSCLGLGTWVTFGSQISDETAEDVLTVAYEHGVNLFDTAEVYAAGKAERTLGNILKSKGWRRSSYVITTKIFWGGQAETERGLSRKHIIEGLRGSLERLQLGYVDIVFANRSDPNCPMEEIVRAMTYVINQGLALYWGTSRWGAAEIMEAYSMARQFNLIPPVCEQAEHHLFQREKVEMQLPELYHKIGVGSVTWYPLACGLITSKYDGRVPDTCRASIKGYQWLKDKVQSEDGKKQQAKVMDLLPVAHQLGCTVAQLAIAWCLRSEGVSSVLLGVSSAEQLIEHLGALQVLSQLTPQTVMEIDGLLGNKPHSKK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AKR6A9/KCNA3.1B/KCNA3B/KV-BETA-3/MGC116886
Common Name KCNAB3
Gene Symbol KCNAB3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.