Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ketohexokinase Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $409.50
Specifications
Antigen | Ketohexokinase |
---|---|
Clone | 1020613 |
Dilution | Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8177SP
![]() |
R&D Systems
MAB8177SP |
25 μg |
Each for $200.50
|
|
|||||
MAB8177100
![]() |
R&D Systems
MAB8177100 |
100 μg |
Each for $409.50
|
|
|||||
Description
Ketohexokinase Monoclonal specifically detects Ketohexokinase in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Ketohexokinase | |
Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
EC 2.7.1.3, Fructokinase, Hepatic fructokinase, ketohexokinase, ketohexokinase (fructokinase), KHK | |
E. coli-derived human Ketohexokinase, Met1-Val298, Accession # P50053 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1020613 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P50053 | |
3795 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title