Learn More
Human KGF/FGF-7 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | KGF/FGF-7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 29568 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P21781 |
| Gene Alias | FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor) |
| Gene Symbols | FGF7 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.