Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KGF/FGF-7 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF251
This item is not returnable.
View return policy
Description
KGF/FGF-7 Polyclonal specifically detects KGF/FGF-7 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
KGF/FGF-7 | |
Polyclonal | |
Biotin | |
Q6FGV5 | |
FGF7 | |
E. coli-derived recombinant human KGF/FGF-7 (R&D Systems, Catalog # 251-KG) Cys32-Thr194 Accession # Q6FGV5 | |
50 μg | |
Primary | |
Detects human KGF/FGF-7 in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human (rh) FGF acidic, rhFGF basic, rhFGF-4, rhFGF-5, and rhFGF-6 is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor) | |
Goat | |
Affinity Purified | |
RUO | |
2252 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction