Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ki67/MKI67 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC7617T100UG
This item is not returnable.
View return policy
Description
Ki67/MKI67 Monoclonal specifically detects Ki67/MKI67 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Ki67/MKI67 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, MKI67, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105, TSG126 | |
Human Ki-67/MKI67 synthetic peptide Accession # P46013 | |
100 μg | |
4288 | |
Human | |
Supernatant |
Flow Cytometry | |
1297A | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P46013 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human Ki-67/MKI67 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction