Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ki67/MKI67 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $538.00
Specifications
Antigen | Ki67/MKI67 |
---|---|
Clone | 1297A |
Dilution | Simple Western 20 ug/mL, Immunohistochemistry 3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 0.3-25 ug/mL, Knockout Validated |
Applications | Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunoassay |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB7617SP
![]() |
R&D Systems
MAB7617SP |
25 μg |
Each for $214.00
|
|
|||||
MAB7617100
![]() |
R&D Systems
MAB7617100 |
100 μg |
Each for $538.00
|
|
|||||
Description
Ki67/MKI67 Monoclonal specifically detects Ki67/MKI67 in Human samples. It is validated for Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, Knockout Validated.Specifications
Ki67/MKI67 | |
Simple Western 20 ug/mL, Immunohistochemistry 3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 0.3-25 ug/mL, Knockout Validated | |
Monoclonal | |
Rabbit | |
Human | |
P46013 | |
4288 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
1297A | |
Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunoassay | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, MKI67, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105, TSG126 | |
Human/Mouse Ki67/MKI67 synthetic peptide, Accession # E9PVX6 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Ki-67/MKI67 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title